Gene Transcripts

Gene Symbol
Myl12b
Entrez Gene ID
50685
Gene Name
myosin, light chain 12B, regulatory
Gene Aliases
Mrlc2, Mrlcb, Mylc2b
Chromosome Location
Chr. 9: 119307168 - 119321500 on Build Rnor_6.0
UniGene ID
Rn.103179
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017343.2 NP_059039.2 2 216
GenBank mRNA BC060577.1 - 2 215
CF977112.1 - 2 220
D14688.1 - 2 186
EV769828.1 - 2 203
FQ215510.1 - 2 202
FQ229195.1 - 2 202
X52840.1 - 1 133

Gene Symbol
Myl12a
Entrez Gene ID
501203
Gene Name
myosin, light chain 12A, regulatory, non-sarcomeric
Gene Aliases
RGD1309537, Rlc-a
Chromosome Location
Chr. 9: 119325282 - 119332967 on Build Rnor_6.0
UniGene ID
Rn.103179
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135017.1 NP_001128489.1 2 243
XM_006245667.2 XP_006245729.1 2 397
XM_006245668.2 XP_006245730.1 2 338
GenBank mRNA EV767772.1 - 273
FQ215121.1 - 2 198
FQ215776.1 - 387
FQ217558.1 - 2 173
FQ220614.1 - 229
FQ223132.1 - 2 207
FQ228530.1 - 2 200
FQ229140.1 - 2 452
FQ232936.1 - 2 179
X05566.1 - 2 185
Z32519.1 - 2 185

View Full Product Details