Gene Transcripts

Gene Symbol
Shisa3
Entrez Gene ID
330096
Gene Name
shisa family member 3
Gene Aliases
D830007B15Rik
Chromosome Location
Chr. 5: 67607883 - 67613987 on Build GRCm38
UniGene ID
Mm.442747
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033415.3 NP_001028587.1 2 3407
GenBank mRNA AK077061.1 - 3 2558

View Full Product Details