Gene Transcripts

Gene Symbol
Mageb4
Entrez Gene ID
434903
Gene Name
melanoma antigen, family B, 4
Gene Aliases
CN716893, Mage-b4, mMage-b4
Chromosome Location
Chr. X: 86250256 - 86256219 on Build GRCm38
UniGene ID
Mm.213584
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033492.2 NP_001028664.2 2137
GenBank mRNA AK144342.1 - 2141
AK144350.1 - 2106
AK165797.1 - 2121
BB807197.1 - 1 99

View Full Product Details