Gene Transcripts

Gene Symbol
Rufy4
Entrez Gene ID
435626
Gene Name
RUN and FYVE domain containing 4
Gene Aliases
F930048N03Rik
Chromosome Location
Chr. 1: 74124618 - 74148230 on Build GRCm38
UniGene ID
Mm.240447
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001034060.3 NP_001029232.2 8 988
NM_001170641.1 NP_001164112.1 8 1011
GenBank mRNA AK158260.1 - 8 993
BC150907.1 - 8 979 9087818
BC150910.1 - 8 979 9087821

View Full Product Details