Gene Transcripts

Gene Symbol
Rgs11
Entrez Gene ID
50782
Gene Name
regulator of G-protein signaling 11
Gene Aliases
C78048
Chromosome Location
Chr. 17: 26202962 - 26211324 on Build GRCm38
UniGene ID
Mm.109840
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081069.1 NP_001074538.1 15 1110
GenBank mRNA AF061934.1 - 14 1015
AK039314.1 - 14 1732
AK080707.1 - 15 1204
AK161551.1 - 11 1169
AK166716.1 - 1 355
BC019741.1 - 4 442
BC049910.1 - 13 1814

View Full Product Details