Gene Transcripts

Gene Symbol
Spag11b
Entrez Gene ID
546038
Gene Name
sperm associated antigen 11B
Gene Aliases
EG546038, Ep2c/h, Spag11c/h
Chromosome Location
Chr. 8: 19140759 - 19143010 on Build GRCm38
UniGene ID
Mm.99206
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001034905.2 NP_001030077.1 2 1385
NM_001039563.3 NP_001034652.2 3 287
NM_001286493.1 NP_001273422.1 3 290
GenBank mRNA AK136627.1 - 2 1386
AY552530.1 - 3 253
BC145547.1 - 3 287 9054111
BC145549.1 - 3 284 9054113
BC145818.1 - 3 284 40131121
BC145820.1 - 3 284 40131123
DQ012055.1 - 3 248

View Full Product Details