Gene Transcripts

Gene Symbol
Pydc4
Entrez Gene ID
623121
Gene Name
pyrin domain containing 4
Gene Aliases
E030037K03Rik, E430029J22Rik
Chromosome Location
Chr. 1: 173566595 - 173599302 on Build GRCm38
UniGene ID
Mm.138449
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001177349.1 NP_001170820.1 4 1469
NM_001177350.1 NP_001170821.1 3 1417
GenBank mRNA AK143015.1 - 4 1470

View Full Product Details