Gene Transcripts

Gene Symbol
Vmn2r92
Entrez Gene ID
627111
Gene Name
vomeronasal 2, receptor 92
Gene Aliases
EG627111
Chromosome Location
Chr. 17: 18151930 - 18185178 on Build GRCm38
UniGene ID
Mm.469863
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001104541.1 NP_001098011.1 1 195

View Full Product Details