Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_025554.2 | NP_079830.2 | 2 | 118 | |
GenBank mRNA | AK010567.1 | - | 2 | 126 | |
AK015823.1 | - | 2 | 116 | ||
AK088610.1 | - | 2 | 174 | ||
AK132109.1 | - | 2 | 173 | ||
AK144668.1 | - | 2 | 179 | ||
AK145201.1 | - | 2 | 182 | ||
AK155368.1 | - | 2 | 151 | ||
BC005599.1 | - | 2 | 171 | ||
BC026842.1 | - | 2 | 109 | 4210827 | |
BC037681.1 | - | 2 | 173 | ||
BC045521.1 | - | 2 | 118 | 2647754 |