Gene Transcripts
- Gene Symbol
- Rhobtb1
- Entrez Gene ID
- 69288
- Gene Name
- Rho-related BTB domain containing 1
- Gene Aliases
- 1700008H16Rik, 3110048G13Rik, AI173445, AI573858, AV350930
- Chromosome Location
- Chr. 10: 69151279 - 69291791 on Build GRCm38
- UniGene ID
- Mm.26659
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001081347.1 |
NP_001074816.1
|
2, 3 |
140 |
|
NM_001252636.1 |
NP_001239565.1
|
2, 3 |
123 |
|
NM_001252637.1 |
NP_001239566.1
|
2, 3 |
244 |
|
NM_001252638.1 |
NP_001239567.1
|
2, 3 |
244 |
|
GenBank mRNA |
AK005770.1 |
- |
2, 3 |
123 |
|
AK034826.1 |
- |
2, 3 |
140 |
|
AK078306.1 |
- |
1 |
18 |
|
BB611014.1 |
- |
2, 3 |
246 |
|
BC099973.1 |
- |
2, 3 |
121 |
6432524 |
BC137646.1 |
- |
1, 2 |
74 |
8860665 |
BC137647.1 |
- |
1, 2 |
74 |
8860666 |
BC144800.1 |
- |
1, 2 |
74 |
9053348 |
View Full Product Details