Gene Transcripts

Gene Symbol
Mrgbp
Entrez Gene ID
73247
Gene Name
MRG/MORF4L binding protein
Gene Aliases
1600027N09Rik, AW060503, C80444
Chromosome Location
Chr. 2: 180581222 - 180586304 on Build GRCm38
UniGene ID
Mm.426558
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028479.1 NP_082755.1 1 194
GenBank mRNA BC116732.1 - 1 194
BC116734.1 - 1 194 40126053

View Full Product Details