Gene Transcripts

Gene Symbol
Ube2ql1
Entrez Gene ID
76980
Gene Name
ubiquitin-conjugating enzyme E2Q family-like 1
Gene Aliases
3110006E14Rik
Chromosome Location
Chr. 13: 69702832 - 69739897 on Build GRCm38
UniGene ID
Mm.23960
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145162.1 NP_001138634.1 2 1753
GenBank mRNA AK014009.1 - 1 186
BC058543.1 - 2 688

View Full Product Details