Gene Transcripts

Gene Symbol
H2afv
Entrez Gene ID
77605
Gene Name
H2A histone family, member V
Gene Aliases
C530002L11Rik, H2a.z-2, H2av
Chromosome Location
Chr. 11: 6427226 - 6444443 on Build GRCm38
UniGene ID
Mm.27624
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029938.1 NP_084214.1 5 556
GenBank mRNA AK021267.1 - 5 479
BC028539.1 - 4 351

View Full Product Details