Gene Transcripts
- Gene Symbol
- Sult1d1
- Entrez Gene ID
- 60393
- Gene Name
- sulfotransferase family 1D, member 1
- Gene Aliases
- ST1D1, Sult-n, Sultn
- Chromosome Location
- Chr. 14: 22107418 - 22126011 on Build Rnor_6.0
- UniGene ID
- Rn.45325
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_021769.1 |
NP_068537.1
|
7 |
816 |
|
GenBank mRNA |
U32372.1 |
- |
7 |
816 |
|
View Full Product Details