Gene Transcripts

Gene Symbol
Mlst8
Entrez Gene ID
64226
Gene Name
MTOR associated protein, LST8 homolog (S. cerevisiae)
Gene Aliases
Gbl
Chromosome Location
Chr. 10: 13839250 - 13845001 on Build Rnor_6.0
UniGene ID
Rn.9444
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022404.2 NP_071799.2 2, 3 501
GenBank mRNA AF051155.2 - 2, 3 500
BC097319.1 - 2, 3 179 7455095

View Full Product Details