Gene Transcripts
- Gene Symbol
- Nr1i3
- Entrez Gene ID
- 65035
- Gene Name
- nuclear receptor subfamily 1, group I, member 3
- Gene Aliases
- CAR
- Chromosome Location
- Chr. 13: 89586224 - 89591278 on Build Rnor_6.0
- UniGene ID
- Rn.163229
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001270838.1 |
NP_001257767.1
|
2, 3 |
284 |
|
NM_001270839.1 |
NP_001257768.1
|
2, 3 |
284 |
|
NM_001270840.1 |
NP_001257769.1
|
2, 3 |
284 |
|
NM_022941.4 |
NP_075230.1
|
2, 3 |
284 |
|
XM_006250261.2 |
XP_006250323.1
|
2, 3 |
393 |
|
XM_008769758.1 |
XP_008767980.1
|
2, 3 |
396 |
|
XM_008769759.1 |
XP_008767981.1
|
2, 3 |
396 |
|
GenBank mRNA |
AB104736.1 |
- |
2, 3 |
337 |
|
AB105072.1 |
- |
2, 3 |
337 |
|
AB204897.1 |
- |
1, 2 |
135 |
|
AB204898.1 |
- |
1, 2 |
135 |
|
AB204899.1 |
- |
1, 2 |
135 |
|
AB204900.1 |
- |
1, 2 |
135 |
|
AF133094.1 |
- |
1, 2 |
135 |
|
AF133095.1 |
- |
1, 2 |
135 |
|
BC089763.1 |
- |
2, 3 |
284 |
7324318 |
View Full Product Details