Gene Transcripts

Gene Symbol
Scamp5
Entrez Gene ID
65171
Gene Name
secretory carrier membrane protein 5
Gene Aliases
Chromosome Location
Chr. 8: 62222648 - 62248011 on Build Rnor_6.0
UniGene ID
Rn.24420
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031726.1 NP_113914.1 5, 6 559
GenBank mRNA AF240784.1 - 5, 6 559

View Full Product Details