Gene Transcripts

Gene Symbol
Rbbp7
Entrez Gene ID
83712
Gene Name
retinoblastoma binding protein 7
Gene Aliases
Chromosome Location
Chr. X: 33648682 - 33665821 on Build Rnor_6.0
UniGene ID
Rn.3600
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031816.1 NP_114004.1 3 450
GenBank mRNA AF090306.1 - 3 450
BC062012.1 - 3 359 5621292

View Full Product Details