Gene Transcripts
- Gene Symbol
- Scg3
- Entrez Gene ID
- 116635
- Gene Name
- secretogranin III
- Gene Aliases
- SgIII
- Chromosome Location
- Chr. 8: 82582352 - 82641536 on Build Rnor_6.0
- UniGene ID
- Rn.11093
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_053856.2 |
NP_446308.1
|
1, 2 |
383 |
|
GenBank mRNA |
U02983.1 |
- |
1, 2 |
385 |
|
View Full Product Details