Gene Transcripts

Gene Symbol
Hdgfl1
Entrez Gene ID
171074
Gene Name
hepatoma derived growth factor-like 1
Gene Aliases
Hdgfrp1, Pwwp1
Chromosome Location
Chr. 17: 40170522 - 40172518 on Build Rnor_6.0
UniGene ID
Rn.81074
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133549.2 NP_598233.2 1 232
GenBank mRNA AF458586.1 - 1 202
AY061636.1 - 1 214
BC085707.1 - 1 232 7132556

View Full Product Details