Gene Transcripts

Gene Symbol
Pbld1
Entrez Gene ID
171564
Gene Name
phenazine biosynthesis-like protein domain containing 1
Gene Aliases
Mawbp, Pbld
Chromosome Location
Chr. 20: 27103300 - 27118652 on Build Rnor_6.0
UniGene ID
Rn.2417
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138530.2 NP_612539.2 402
XM_006256372.2 XP_006256434.1 5, 6 717
GenBank mRNA AY083160.1 - 414
BC078735.1 - 402 7126368
FQ209640.1 - 437
FQ219037.1 - 437

View Full Product Details