Gene Transcripts

Gene Symbol
Tpcn1
Entrez Gene ID
246215
Gene Name
two pore segment channel 1
Gene Aliases
Chromosome Location
Chr. 12: 41507784 - 41566847 on Build Rnor_6.0
UniGene ID
Rn.24484
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_139332.3 NP_647548.2 505
GenBank mRNA AB018253.2 - 508
BC062072.1 - 505 5599545

View Full Product Details