Gene Transcripts

Gene Symbol
TAS2R1
Entrez Gene ID
50834
Gene Name
taste 2 receptor member 1
Gene Aliases
T2R1, TRB7
Chromosome Location
Chr. 5: 9628997 - 9630351 on Build GRCh38
UniGene ID
Hs.567492
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019599.2 NP_062545.1 1 772
GenBank mRNA BC101729.1 - 1 526
BC101731.1 - 1 526 8069237

View Full Product Details