Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001177647.2 | NP_001171118.1 | 6 | 1076 | |
NM_001291019.1 | NP_001277948.1 | 8 | 1742 | ||
NM_001291020.1 | NP_001277949.1 | 9 | 1544 | ||
NM_001291021.1 | NP_001277950.1 | 6 | 1088 | ||
NM_001291022.1 | NP_001277951.1 | 5 | 719 | ||
NM_007547.4 | NP_031573.2 | 8 | 1730 | ||
GenBank mRNA | AB018194.1 | - | 8 | 1296 | |
AF072543.1 | - | 8 | 1286 | ||
AF072544.1 | - | 6 | 632 | ||
AF332079.1 | - | 8 | 1296 | ||
AF332080.1 | - | 8 | 1296 | ||
AK159617.1 | - | 9 | 1551 | ||
BC062197.1 | - | 8 | 1321 | 5368250 | |
CJ151068.1 | - | 5 | 320 | ||
CX236057.1 | - | 6 | 673 | ||
D85785.1 | - | 8 | 1580 | ||
D87967.1 | - | 8 | 1695 | ||
D87968.1 | - | 8 | 1707 | ||
U89694.1 | - | 8 | 1307 | ||
Y10349.1 | - | 8 | 1353 |