Gene Transcripts
- Gene Symbol
- Drg1
- Entrez Gene ID
- 13494
- Gene Name
- developmentally regulated GTP binding protein 1
- Gene Aliases
- AA408859, AI132520, Nedd3
- Chromosome Location
- Chr. 11: 3249922 - 3266386 on Build GRCm38
- UniGene ID
- Mm.3250
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_007879.1 |
NP_031905.1
|
2, 3 |
244 |
|
GenBank mRNA |
AK044194.1 |
- |
2, 3 |
253 |
|
AK080814.1 |
- |
2, 3 |
242 |
|
AK090156.1 |
- |
2, 3 |
274 |
|
AK144438.1 |
- |
2, 3 |
246 |
|
AK155061.1 |
- |
2, 3 |
259 |
|
AK208688.1 |
- |
1, 2 |
120 |
|
AK210426.1 |
- |
1, 2 |
19 |
|
AK211821.1 |
- |
1, 2 |
19 |
|
BC039649.1 |
- |
2, 3 |
238 |
5351885 |
D10715.1 |
- |
2, 3 |
244 |
|
View Full Product Details