Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_008496.4 | NP_032522.2 | 2, 3 | 198 | |
GenBank mRNA | AF038562.1 | - | 123 | ||
AF331640.1 | - | 1, 2 | 103 | ||
AK003153.1 | - | 2, 3 | 132 | ||
AK008773.1 | - | 2, 3 | 132 | ||
AK009151.1 | - | 2, 3 | 141 | ||
BC116838.1 | - | 2, 3 | 159 | 40126157 | |
BC116840.1 | - | 2, 3 | 159 | 40126159 | |
BG803457.1 | - | 2, 3 | 355 |