Gene Transcripts

Gene Symbol
Sh2b3
Entrez Gene ID
16923
Gene Name
SH2B adaptor protein 3
Gene Aliases
AI429800, Lnk
Chromosome Location
Chr. 5: 121815481 - 121837655 on Build GRCm38
UniGene ID
Mm.65493
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK028796.1 - 7 1546
AK156499.1 - 5 1505
AK157297.1 - 6 1415
AK165124.1 - 4 735
AK171128.1 - 7 1489
AK172573.1 - 6 1419
AK187693.1 - 15
BC006759.1 - 6 1399 3709861
U89992.2 - 1310

View Full Product Details