Gene Transcripts

Gene Symbol
Mut
Entrez Gene ID
17850
Gene Name
methylmalonyl-Coenzyme A mutase
Gene Aliases
D230010K02Rik, Mcm
Chromosome Location
Chr. 17: 40934685 - 40961989 on Build GRCm38
UniGene ID
Mm.259884
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008650.3 NP_032676.2 2 308
GenBank mRNA AK146309.1 - 2 268
BC019175.1 - 2 290 5007694
BY262299.1 - 2 309
X51941.1 - 1 219

View Full Product Details