Gene Transcripts
- Gene Symbol
- Scg3
- Entrez Gene ID
- 20255
- Gene Name
- secretogranin III
- Gene Aliases
- 1B1075, AI385542, Chgd, SgIII
- Chromosome Location
- Chr. 9: 75643366 - 75684082 on Build GRCm38
- UniGene ID
- Mm.2386
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001164790.1 |
NP_001158262.1
|
1, 2 |
421 |
|
NM_009130.3 |
NP_033156.1
|
1, 2 |
421 |
|
GenBank mRNA |
AK133655.1 |
- |
1, 2 |
323 |
|
AK139140.1 |
- |
1, 2 |
361 |
|
AK148498.1 |
- |
1, 2 |
403 |
|
AK158315.1 |
- |
1, 2 |
294 |
|
AK164089.1 |
- |
1, 2 |
364 |
|
BC024785.1 |
- |
1, 2 |
380 |
5362975 |
BU058741.1 |
- |
1, 2 |
421 |
|
U02982.1 |
- |
1, 2 |
385 |
|
View Full Product Details