Gene Transcripts

Gene Symbol
Sap18
Entrez Gene ID
20220
Gene Name
Sin3-associated polypeptide 18
Gene Aliases
C530046K05Rik, D11Ertd539e, EMegR4, Sinbp1
Chromosome Location
Chr. 14: 57798189 - 57804980 on Build GRCm38
UniGene ID
Mm.30173
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009119.3 NP_033145.2 3 351
GenBank mRNA AK005189.1 - 3 269
AK006647.1 - 3 290
AK085992.1 - 3 290
AK133619.1 - 3 285
AK144904.1 - 3 324
AK146928.1 - 3 324
AK209620.1 - 2 108
BC006625.1 - 3 292 3968600
BC099480.1 - 3 285 30934194
CF730347.1 - 3 351
Z97062.1 - 3 286

Gene Symbol
Gm10094
Entrez Gene ID
100041953
Gene Name
predicted gene 10094
Gene Aliases
ENSMUSG00000061104
Chromosome Location
Chr. 8: 95825356 - 95826156 on Build GRCm38
UniGene ID
Mm.290178
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001142441.1 NP_001135913.1 1 322

View Full Product Details