Gene Transcripts

Gene Symbol
PDZK1IP1
Entrez Gene ID
10158
Gene Name
PDZK1 interacting protein 1
Gene Aliases
DD96, MAP17, SPAP
Chromosome Location
Chr. 1: 47183589 - 47190099 on Build GRCh38
UniGene ID
Hs.431099
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005764.3 NP_005755.1 3, 4 447
GenBank mRNA BC012303.1 - 3, 4 360
BI763294.1 - 1, 2 112
BM924665.1 - 3, 4 473
CR450304.1 - 3, 4 280

View Full Product Details