Gene Transcripts

Gene Symbol
Tll1
Entrez Gene ID
21892
Gene Name
tolloid-like
Gene Aliases
Tll, Tll-1
Chromosome Location
Chr. 8: 64014150 - 64205993 on Build GRCm38
UniGene ID
Mm.418367
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009390.2 NP_033416.2 2 848
GenBank mRNA AK080919.1 - 2 864
U34042.1 - 2 848

View Full Product Details