Gene Transcripts
- Gene Symbol
- Ulk1
- Entrez Gene ID
- 22241
- Gene Name
- unc-51 like kinase 1
- Gene Aliases
- AU041434, Unc51.1, mKIAA0722
- Chromosome Location
- Chr. 5: 110784489 - 110810118 on Build GRCm38
- UniGene ID
- Mm.271898
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_009469.3 |
NP_033495.2
|
2, 3 |
569 |
|
GenBank mRNA |
AF053756.1 |
- |
2, 3 |
261 |
|
AF072370.1 |
- |
2, 3 |
252 |
|
AK122357.1 |
- |
1, 2 |
71 |
|
AK148317.1 |
- |
2, 3 |
544 |
|
AK154939.1 |
- |
2, 3 |
586 |
|
AK156534.1 |
- |
2, 3 |
546 |
|
BC053062.1 |
- |
2, 3 |
229 |
|
BC057121.1 |
- |
2, 3 |
569 |
6834534 |
BC059835.1 |
- |
2, 3 |
520 |
6406755 |
View Full Product Details