Gene Transcripts

Gene Symbol
Man2b1
Entrez Gene ID
17159
Gene Name
mannosidase 2, alpha B1
Gene Aliases
AW107687, LAMAN
Chromosome Location
Chr. 8: 85082729 - 85098739 on Build GRCm38
UniGene ID
Mm.4219
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010764.2 NP_034894.2 7 982
GenBank mRNA AK004817.2 - 7 939
AK035551.1 - 7 937
AK054315.1 - 4 2272
AK147928.1 - 7 940
AK150154.1 - 4 314
AK170848.1 - 4 313
AK171166.1 - 7 940
AK171377.1 - 4 313
BC005430.1 - 7 927 3488681
BC052517.1 - 2 881
U29947.1 - 7 925
U87240.1 - 7 916

View Full Product Details