Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_010950.2 | NP_035080.2 | 6 | 650 | |
GenBank mRNA | AK147523.1 | - | 4 | 6437 | |
AK196401.1 | - | 3 | 183 | ||
BC059915.1 | - | 5 | 1239 | ||
BC068116.1 | - | 6 | 477 | 6809443 | |
BC098097.1 | - | 6 | 491 | 6403012 | |
U96441.1 | - | 6 | 534 |