Gene Transcripts

Gene Symbol
RAD17
Entrez Gene ID
5884
Gene Name
RAD17 checkpoint clamp loader component
Gene Aliases
CCYC, HRAD17, R24L, RAD17SP, RAD24
Chromosome Location
Chr. 5: 69369297 - 69414801 on Build GRCh38
UniGene ID
Hs.16184
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001278622.1 NP_001265551.1 9 1318
NM_002873.1 NP_002864.1 8 1423
NM_133338.2 NP_579916.1 10 1319
NM_133339.2 NP_579917.1 7 876
NM_133340.2 NP_579918.1 7 909
NM_133341.2 NP_579919.1 7 835
NM_133342.2 NP_579920.1 9 1296
NM_133343.1 NP_579921.1 9 1261
NM_133344.2 NP_579922.1 8 1078
XM_017009679.1 XP_016865168.1 9 1138
XM_017009680.1 XP_016865169.1 8 1192
XM_017009681.1 XP_016865170.1 8 1050
GenBank mRNA AF017748.1 - 8 1048
AF076838.1 - 8 1423
AF085736.2 - 7 876
AF098533.1 - 7 855
AF098534.1 - 7 781
AF112263.1 - 7 900
AF126424.1 - 10 1318
AJ001642.1 - 8 1087
AJ004977.1 - 8 1083
AK292487.1 - 9 1296
AL122068.1 - 9 1261
BC032304.1 - 7 889 5170876
BX537441.1 - 9 1318

View Full Product Details