Gene Transcripts

Gene Symbol
Sbp
Entrez Gene ID
20234
Gene Name
spermine binding protein
Gene Aliases
AV082137, AV082259, p25
Chromosome Location
Chr. 17: 23941996 - 23945607 on Build GRCm38
UniGene ID
Mm.390958
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011321.3 NP_035451.1 3 574
GenBank mRNA AK035423.1 - 3 575
BC049605.1 - 3 574 6743567
X06246.1 - 3 574

View Full Product Details