Gene Transcripts
- Gene Symbol
- Stk11
- Entrez Gene ID
- 20869
- Gene Name
- serine/threonine kinase 11
- Gene Aliases
- AA408040, Lkb1, Par-4, R75140
- Chromosome Location
- Chr. 10: 80115693 - 80130685 on Build GRCm38
- UniGene ID
- Mm.44231
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AB015801.1 |
- |
2, 3 |
407 |
|
AF129870.1 |
- |
2, 3 |
785 |
|
AF145287.1 |
- |
2, 3 |
389 |
|
AF151711.1 |
- |
2, 3 |
389 |
|
AK147802.1 |
- |
2, 3 |
448 |
|
AK171909.1 |
- |
3, 4 |
1183 |
|
AK172385.1 |
- |
2, 3 |
1172 |
|
AK172528.1 |
- |
2, 3 |
1169 |
|
BC052379.1 |
- |
2, 3 |
450 |
5688695 |
CO424479.1 |
- |
2, 3 |
607 |
|
EU730638.1 |
- |
2, 3 |
368 |
|
View Full Product Details