Gene Transcripts

Gene Symbol
Sprr3
Entrez Gene ID
20766
Gene Name
small proline-rich protein 3
Gene Aliases
SPR3
Chromosome Location
Chr. 3: 92456502 - 92458720 on Build GRCm38
UniGene ID
Mm.57092
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001204427.1 NP_001191356.1 2 256
NM_011478.2 NP_035608.1 2 392
GenBank mRNA BC125629.1 - 1 286
BY709657.1 - 2 392

View Full Product Details