Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001040435.2 | NP_001035525.1 | 12 | 2048 | |
GenBank mRNA | AF093542.1 | - | 13 | 1282 | |
AF156934.1 | - | 12 | 1602 | ||
AF203445.1 | - | 12 | 1549 | ||
AF203446.1 | - | 12 | 937 | ||
AF247674.1 | - | 13 | 1333 | ||
AK076282.1 | - | 2075 | |||
AK145556.1 | - | 12 | 1609 | ||
AK160291.1 | - | 12 | 1611 | ||
AK183147.1 | - | 4 | 262 | ||
BC003252.1 | - | 12 | 1605 | 3500413 | |
BC055071.1 | - | 4024 | |||
BC066184.1 | - | 12 | 1584 | 30457053 | |
BC068314.1 | - | 12 | 1584 | 30461783 |