Gene Transcripts

Gene Symbol
Cln8
Entrez Gene ID
26889
Gene Name
ceroid-lipofuscinosis, neuronal 8
Gene Aliases
mnd
Chromosome Location
Chr. 8: 14881335 - 14901720 on Build GRCm38
UniGene ID
Mm.254027
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012000.3 NP_036130.1 3 2666
GenBank mRNA AK049145.1 - 3 2339
AK089692.1 - 4 2107

View Full Product Details