Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_013591.2 | NP_038619.2 | 3 | 545 | |
GenBank mRNA | AK137028.1 | - | 3 | 546 | |
AK156096.1 | - | 3 | 523 | ||
BC024372.1 | - | 3 | 538 | 4986802 | |
BC115910.1 | - | 3 | 539 | 40105157 | |
BC118056.1 | - | 3 | 539 | 40105156 | |
L21203.1 | - | 3 | 542 | ||
S80258.1 | - | 3 | 512 |