Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001083319.1 | NP_001076788.1 | 13 | 1628 | |
NM_013699.2 | NP_038727.1 | 12 | 1520 | ||
GenBank mRNA | AB221581.1 | - | 11 | 1108 | |
AK079815.1 | - | 5 | 355 | ||
AK140942.1 | - | 7 | 525 | ||
AK146040.1 | - | 1 | 140 | ||
AY182062.1 | - | 12 | 1217 | ||
BC034749.1 | - | 11 | 1847 | ||
BC139828.1 | - | 11 | 1137 | 40142981 | |
BC141082.1 | - | 11 | 1199 | 9055694 | |
U20086.1 | - | 11 | 1111 |