Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_013768.3 | NP_038796.2 | 2 | 277 | |
GenBank mRNA | AF167573.1 | - | 2 | 145 | |
AK076346.1 | - | 2 | 168 | ||
AK084106.1 | - | 2 | 278 | ||
AK132414.1 | - | 2 | 169 | ||
AK165165.1 | - | 2 | 195 | ||
BC023905.1 | - | 2 | 224 | 5321370 | |
BY064945.1 | - | 2 | 333 |