Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_013781.3 | NP_038809.1 | 2 | 274 | |
GenBank mRNA | AB043953.1 | - | 2 | 257 | |
AF168364.1 | - | 2 | 215 | ||
AK155165.1 | - | 2 | 210 | ||
AK172072.1 | - | 2 | 195 | ||
BC113203.1 | - | 2 | 229 | 40048064 |