Gene Transcripts

Gene Symbol
Tcl1b1
Entrez Gene ID
27379
Gene Name
T cell leukemia/lymphoma 1B, 1
Gene Aliases
C88214
Chromosome Location
Chr. 12: 105159704 - 105166625 on Build GRCm38
UniGene ID
Mm.241933
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013773.2 NP_038801.1 1, 2 208
GenBank mRNA AF195488.1 - 1, 2 208
AK136181.1 - 1, 2 224
BC052337.1 - 1, 2 244 30053367

View Full Product Details