Gene Transcripts

Gene Symbol
Sh3bgr
Entrez Gene ID
50795
Gene Name
SH3-binding domain glutamic acid-rich protein
Gene Aliases
5430437A18Rik
Chromosome Location
Chr. 16: 96200470 - 96228937 on Build GRCm38
UniGene ID
Mm.28638
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015825.2 NP_056640.1 4, 5 626
GenBank mRNA AJ239082.1 - 4, 5 392
AJ272170.1 - 4, 5 454
BC055870.1 - 4, 5 443

View Full Product Details