Gene Transcripts

Gene Symbol
Sall2
Entrez Gene ID
50524
Gene Name
sal-like 2 (Drosophila)
Gene Aliases
AI225809, AW559097, Msal-2, Sal-2, mKIAA0360, p150(Sal2)
Chromosome Location
Chr. 14: 52311177 - 52328670 on Build GRCm38
UniGene ID
Mm.39487
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001244916.1 NP_001231845.1 2 3451
NM_015772.3 NP_056587.2 2 3738
GenBank mRNA AB093233.1 - 2 3397
AJ007396.1 - 2 3829
AK077601.1 - 2 3365
BC054527.1 - 2 3432
BC085361.1 - 2 3515 5706710

View Full Product Details