Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_019662.2 | NP_062636.2 | 4 | 793 | |
GenBank mRNA | AF084466.1 | - | 3 | 536 | |
AK077271.1 | - | 4 | 791 | ||
AK087473.1 | - | 4 | 766 | ||
AK159550.1 | - | 3 | 576 | ||
BC057138.1 | - | 4 | 772 | 5149047 |