Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001167910.1 | NP_001161382.1 | 3 | 753 | |
NM_019709.4 | NP_062683.3 | 4 | 902 | ||
GenBank mRNA | AF094820.1 | - | 2 | 321 | |
AK029048.1 | - | 3 | 730 | ||
AK122202.1 | - | 3 | 719 | ||
AK147336.1 | - | 3 | 698 | ||
AK147495.1 | - | 3 | 727 | ||
BC011533.1 | - | 3 | 719 | ||
BC054837.1 | - | 2 | 521 | 6490394 | |
BC057198.1 | - | 4 | 868 | 5310414 | |
DQ913734.1 | - | 2 | 384 |